siRNA Details
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 433 | ||
XM_017001691.1 | XP_016857180.1 | 3 | 338 | |||
GenBank mRNA | AF151042.1 | - | 3 | 417 | ||
AF374386.1 | - | 3 | 351 | |||
AK000319.1 | - | 2 | 154 | |||
BC001959.1 | - | 3 | 404 | 3536977 | ||
BC078177.1 | - | 3 | 385 | 6526331 | ||
BC085018.1 | - | 3 | 403 | 6650445 | ||
DA043474.1 | - | 3 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
