siRNA Details
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 820 | ||
GenBank mRNA | BC031811.1 | - | 1 | 820 | ||
BC104232.1 | - | 1 | 428 | 40032247 | ||
BC104233.1 | - | 1 | 430 | 40032251 | ||
BC133657.1 | - | 1 | 430 |
Gene Symbol: LCE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178348.2 | NP_848125.1 | 1 | 320 |
Gene Symbol: LCE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 305 | ||
NM_178351.3 | NP_848128.1 | 2 | 395 |
Gene Symbol: LCE1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178352.2 | NP_848129.1 | 2 | 375 | ||
GenBank mRNA | BG697954.1 | - | 2 | 375 |
Gene Symbol: LCE1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178353.1 | NP_848130.1 | 2 | 397 | ||
GenBank mRNA | BC038391.1 | - | 2 | 397 |
Gene Symbol: LCE1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178354.2 | NP_848131.1 | 1 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
