siRNA Details
Gene Symbol: NBPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143987.1 | NP_001137459.1 | 2 | 283 | ||
NM_001143988.1 | NP_001137460.1 | 2 | 283 | |||
XM_005271144.3 | XP_005271201.1 | 2 | 407 | |||
XM_011542012.2 | XP_011540314.1 | 7 | 1736 | |||
XM_011542014.2 | XP_011540316.1 | 2 | 1045 | |||
XM_017002148.1 | XP_016857637.1 | 7 | 871 | |||
GenBank mRNA | BC125161.1 | - | 2 | 244 |
Gene Symbol: NBPF22P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003719.2 | 1 | 387 | |||
GenBank mRNA | AK308274.1 | - | 1 | 371 | ||
BC050328.1 | - | 1 | 387 |
Gene Symbol: NBPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143989.2 | NP_001137461.1 | 2 | 283 | ||
XM_011540763.1 | XP_011539065.1 | 2 | 269 | |||
GenBank mRNA | AK057395.1 | - | 2 | 253 | ||
AK302318.1 | - | 2 | 278 | |||
AK316438.1 | - | 2 | 269 | |||
BC125162.1 | - | 2 | 244 | |||
DC402863.1 | - | 2 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
