siRNA Details
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 4 | 524 | ||
XM_017015367.1 | XP_016870856.1 | 3 | 441 | |||
XM_017015368.1 | XP_016870857.1 | 3 | 424 | |||
XM_017015369.1 | XP_016870858.1 | 4 | 507 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 3 | 435 | |||
GenBank mRNA | AK128231.1 | - | 3 | 435 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 3 | 296 | ||
XM_006712884.3 | XP_006712947.1 | 3 | 720 | |||
XM_006712887.3 | XP_006712950.1 | 2 | 292 | |||
XM_017005467.1 | XP_016860956.1 | 2 | 292 | |||
XM_017005468.1 | XP_016860957.1 | 2 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
