siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278665.1 | NP_001265594.1 | 1, 2 | 185 | ||
NM_203282.3 | NP_975011.3 | 1, 2 | 185 | |||
XM_017027517.1 | XP_016883006.1 | 1, 2 | 613 | |||
GenBank mRNA | AF054180.1 | - | 1, 2 | 171 | ||
AK291286.1 | - | 1, 2 | 152 | |||
BC043147.1 | - | 1, 2 | 157 | 5296940 | ||
BC056243.1 | - | 1, 2 | 161 | |||
BC139749.1 | - | 1, 2 | 116 | 40115903 |
Gene Symbol: ZNF675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138330.2 | NP_612203.2 | 1, 2 | 192 | ||
XM_017026442.1 | XP_016881931.1 | 1, 2 | 197 | |||
XM_017026443.1 | XP_016881932.1 | 1, 2 | 197 | |||
XM_017026444.1 | XP_016881933.1 | 1, 2 | 189 | |||
GenBank mRNA | AK093669.1 | - | 1, 2 | 179 | ||
AK307845.1 | - | 1, 2 | 120 | |||
AY044432.1 | - | 1, 2 | 117 | |||
BC032394.1 | - | 1, 2 | 174 | |||
DA777369.1 | - | 1, 2 | 211 |
Gene Symbol: ZNF726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244038.1 | NP_001230967.1 | 1, 2 | 123 | ||
GenBank mRNA | AB593087.1 | - | 1, 2 | 86 | ||
BC046415.1 | - | 1, 2 | 95 | 4821872 | ||
BE783649.1 | - | 1, 2 | 95 | |||
CN359557.1 | - | 1, 2 | 123 |
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300951.1 | NP_001287880.1 | 1, 2 | 243 | ||
NM_003430.3 | NP_003421.2 | 1, 2 | 243 | |||
XM_011528266.2 | XP_011526568.1 | 1, 2 | 158 | |||
GenBank mRNA | AK291256.1 | - | 1, 2 | 150 | ||
AK300294.1 | - | 1, 2 | 129 | |||
DB068854.1 | - | 1, 2 | 243 | |||
DB219374.1 | - | 1, 2 | 150 | |||
L11672.1 | - | 1, 2 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
