siRNA Details
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256648.1 | NP_001243577.1 | 5 | 771 | ||
NM_001256649.1 | NP_001243578.1 | 5 | 636 | |||
NM_001256650.1 | NP_001243579.1 | 4 | 508 | |||
NM_001256653.1 | NP_001243582.1 | 2 | 249 | |||
NM_003423.3 | NP_003414.2 | 2 | 258 | |||
XM_011528257.2 | XP_011526559.1 | 2 | 180 | |||
XM_011528259.2 | XP_011526561.1 | 2 | 737 | |||
XM_017027207.1 | XP_016882696.1 | 5 | 6249 | |||
XM_017027208.1 | XP_016882697.1 | 5 | 929 | |||
XM_017027209.1 | XP_016882698.1 | 5 | 5982 | |||
XM_017027210.1 | XP_016882699.1 | 4 | 5773 | |||
XM_017027211.1 | XP_016882700.1 | 5 | 5916 | |||
XM_017027212.1 | XP_016882701.1 | 4 | 776 | |||
XM_017027213.1 | XP_016882702.1 | 5 | 6024 | |||
XM_017027214.1 | XP_016882703.1 | 3 | 699 | |||
XM_017027215.1 | XP_016882704.1 | 4 | 538 | |||
XM_017027216.1 | XP_016882705.1 | 3 | 324 | |||
GenBank mRNA | AK291353.1 | - | 5 | 586 | ||
BC006528.2 | - | 2 | 225 | 3687640 | ||
BT009756.1 | - | 1 | 70 | |||
BX471947.1 | - | 2 | 259 | |||
CR933640.1 | - | 5 | 731 | |||
DR002502.1 | - | 2 | 180 | |||
X59244.1 | - | 4 | 407 |
Gene Symbol: ZNF492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020855.2 | NP_065906.1 | 2 | 236 | ||
GenBank mRNA | AB040906.1 | - | 2 | 123 | ||
BP871882.1 | - | 2 | 236 |
Gene Symbol: ZNF626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076675.2 | NP_001070143.1 | 2 | 238 | ||
NM_145297.3 | NP_660340.1 | 2 | 238 | |||
GenBank mRNA | BC007116.1 | - | 2 | 217 | ||
BC107803.1 | - | 2 | 242 | |||
BC150651.1 | - | 2 | 149 | 9057022 | ||
CR749233.1 | - | 2 | 190 |
Gene Symbol: ZNF676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001411.2 | NP_001001411.2 | 1 | 310 | ||
GenBank mRNA | BX281304.1 | - | 1 | 310 |
Gene Symbol: ZNF681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138286.2 | NP_612143.2 | 2 | 230 | ||
GenBank mRNA | AK122869.1 | - | 2 | 234 | ||
BC136346.1 | - | 2 | 109 | 9020333 |
Gene Symbol: ZNF726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244038.1 | NP_001230967.1 | 2 | 201 | ||
GenBank mRNA | AB593087.1 | - | 2 | 164 | ||
BC046415.1 | - | 2 | 173 | 4821872 | ||
BE783649.1 | - | 2 | 173 | |||
BX475164.1 | - | 2 | 209 | |||
CN359557.1 | - | 2 | 201 |
Gene Symbol: ZNF728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267716.1 | NP_001254645.1 | 2 | 266 | ||
GenBank mRNA | DB076533.1 | - | 2 | 245 |
Gene Symbol: ZNF737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159293.1 | NP_001152765.1 | 2 | 268 | ||
XM_005259697.3 | XP_005259754.1 | 2 | 257 | |||
XM_006722597.3 | XP_006722660.1 | 2 | 257 | |||
XM_011527598.2 | XP_011525900.1 | 2 | 257 | |||
XM_011527599.2 | XP_011525901.1 | 2 | 257 | |||
XM_017026116.1 | XP_016881605.1 | 3 | 355 | |||
GenBank mRNA | DA043454.1 | - | 2 | 268 |
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300951.1 | NP_001287880.1 | 2 | 321 | ||
NM_003430.3 | NP_003421.2 | 2 | 321 | |||
XM_011528265.2 | XP_011526567.1 | 2 | 282 | |||
XM_011528266.2 | XP_011526568.1 | 2 | 236 | |||
XM_017027235.1 | XP_016882724.1 | 2 | 275 | |||
GenBank mRNA | AK291256.1 | - | 2 | 228 | ||
AK300294.1 | - | 2 | 207 | |||
DB068854.1 | - | 2 | 321 | |||
DB219374.1 | - | 2 | 228 | |||
L11672.1 | - | 2 | 209 | |||
M61871.1 | - | 1 | 73 |
Gene Symbol: ZNF98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098626.1 | NP_001092096.1 | 2 | 237 | ||
XM_011527705.1 | XP_011526007.1 | 2 | 196 | |||
XM_017026373.1 | XP_016881862.1 | 2 | 519 | |||
GenBank mRNA | BC110576.1 | - | 1 | 27 | 40007052 | |
BC129810.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
