siRNA Details
Gene Symbol: ZNF726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244038.1 | NP_001230967.1 | 3 | 325 | ||
GenBank mRNA | AB593087.1 | - | 3 | 288 | ||
BC046415.1 | - | 3 | 297 | 4821872 | ||
BE783649.1 | - | 3 | 297 | |||
BX475164.1 | - | 3 | 333 | |||
CN359557.1 | - | 3 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
