siRNA Details
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031623.2 | NP_001026794.1 | 3 | 388 | ||
NM_001257273.1 | NP_001244202.1 | 3 | 408 | |||
NM_015555.2 | NP_056370.2 | 3 | 388 | |||
XM_005248994.1 | XP_005249051.1 | 3 | 408 | |||
XM_011514460.1 | XP_011512762.1 | 2 | 147 | |||
XM_011514462.2 | XP_011512764.1 | 3 | 408 | |||
XM_011514463.2 | XP_011512765.1 | 2 | 165 | |||
GenBank mRNA | AB011148.1 | - | 3 | 207 | ||
AK225806.1 | - | 3 | 215 | |||
AK302577.1 | - | 3 | 253 | |||
AY055204.1 | - | 3 | 389 | |||
BC021712.2 | - | 3 | 305 | 4821000 | ||
BC042450.1 | - | 1 | 74 | |||
BC058853.1 | - | 1 | 74 | |||
BC098382.1 | - | 3 | 231 | 5295845 | ||
BX648081.1 | - | 3 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
