siRNA Details
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 3 | 337 | |||
GenBank mRNA | BC052779.1 | - | 3 | 363 | ||
BC064938.1 | - | 3 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
