siRNA Details
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 603 | ||
GenBank mRNA | BC096732.1 | - | 1 | 596 | 40015059 | |
BC098355.1 | - | 1 | 596 | 40015056 | ||
M11026.1 | - | 1 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
