siRNA Details
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128126.2 | NP_001121598.1 | 3 | 550 | ||
NM_001254726.1 | NP_001241655.1 | 3 | 550 | |||
NM_001254727.1 | NP_001241656.1 | 3 | 550 | |||
NM_001254728.1 | NP_001241657.1 | 3 | 907 | |||
NM_001254729.1 | NP_001241658.1 | 3 | 425 | |||
NM_007077.4 | NP_009008.2 | 3 | 550 | |||
XM_005267293.4 | XP_005267350.1 | 4 | 993 | |||
XM_011536371.2 | XP_011534673.1 | 4 | 563 | |||
XM_011536372.2 | XP_011534674.1 | 3 | 957 | |||
GenBank mRNA | AB030654.1 | - | 3 | 370 | ||
AF155159.1 | - | 3 | 344 | |||
AK304115.1 | - | 3 | 371 | |||
AK316517.1 | - | 3 | 373 | |||
BC001259.1 | - | 3 | 333 | 3454117 | ||
BF671561.1 | - | 3 | 423 | |||
BG702035.1 | - | 3 | 559 | |||
BT006701.1 | - | 2 | 161 | |||
BX247969.1 | - | 3 | 352 | |||
BX388185.2 | - | 3 | 336 | |||
CR457100.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
