siRNA Details
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128126.2 | NP_001121598.1 | 3, 4 | 622 | ||
NM_001254726.1 | NP_001241655.1 | 3, 4 | 622 | |||
NM_001254727.1 | NP_001241656.1 | 3, 4 | 622 | |||
NM_001254728.1 | NP_001241657.1 | 3, 4 | 979 | |||
NM_001254729.1 | NP_001241658.1 | 3, 4 | 497 | |||
NM_007077.4 | NP_009008.2 | 3, 4 | 622 | |||
XM_005267293.4 | XP_005267350.1 | 4, 5 | 1065 | |||
XM_011536371.2 | XP_011534673.1 | 4, 5 | 635 | |||
XM_011536372.2 | XP_011534674.1 | 3, 4 | 1029 | |||
GenBank mRNA | AB030654.1 | - | 3, 4 | 442 | ||
AF155159.1 | - | 3, 4 | 416 | |||
AK304115.1 | - | 3, 4 | 443 | |||
AK316517.1 | - | 3, 4 | 445 | |||
BC001259.1 | - | 3, 4 | 405 | 3454117 | ||
BF671561.1 | - | 3, 4 | 495 | |||
BG702035.1 | - | 3, 4 | 631 | |||
BT006701.1 | - | 2, 3 | 233 | |||
BX247969.1 | - | 3, 4 | 424 | |||
BX388185.2 | - | 3, 4 | 408 | |||
CR457100.1 | - | 2, 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
