siRNA Details
Gene Symbol: DEFB105A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152250.2 | NP_689463.1 | 2 | 145 | ||
GenBank mRNA | AB089180.1 | - | 2 | 93 | ||
AF540977.1 | - | 2 | 93 | |||
BC128437.1 | - | 2 | 97 | 40119533 | ||
BC128438.1 | - | 2 | 97 | 40119534 | ||
BC140937.1 | - | 2 | 145 | 9056749 |
Gene Symbol: DEFB105B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040703.2 | NP_001035793.1 | 2 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
