siRNA Details
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 7, 8 | 1451 | ||
XM_011516234.2 | XP_011514536.1 | 7, 8 | 2354 | |||
XM_011516235.2 | XP_011514537.1 | 7, 8 | 2354 | |||
GenBank mRNA | BC056606.1 | - | 7, 8 | 1452 | 4838469 | |
BC100972.2 | - | 2, 3 | 376 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 3, 4 | 945 | ||
XM_017012933.1 | XP_016868422.1 | 3, 4 | 949 |
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 3, 4 | 830 | ||
XM_017012887.1 | XP_016868376.1 | 3, 4 | 830 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 3, 4 | 949 | ||
XM_017012888.1 | XP_016868377.1 | 3, 4 | 949 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 4, 5 | 701 | ||
XM_005250743.4 | XP_005250800.1 | 3, 4 | 556 | |||
XM_011516756.2 | XP_011515058.1 | 3, 4 | 470 |
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 5, 6 | 1150 | ||
XM_011516237.2 | XP_011514539.1 | 5, 6 | 1149 | |||
XM_017012223.1 | XP_016867712.1 | 5, 6 | 1149 | |||
GenBank mRNA | BC100973.1 | - | 2, 3 | 388 | 40005226 | |
DR731150.1 | - | 2, 3 | 294 | |||
JQ023160.1 | - | 4, 5 | 676 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 5, 6 | 1150 | ||
XM_005250093.3 | XP_005250150.1 | 4, 5 | 1718 | |||
XM_011515702.2 | XP_011514004.1 | 4, 5 | 1718 |
Gene Symbol: SPDYE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128076.1 | NP_001121548.1 | 3, 4 | 583 | ||
XM_005256632.4 | XP_005256689.1 | 3, 4 | 1452 | |||
XM_005256633.4 | XP_005256690.1 | 3, 4 | 1452 | |||
GenBank mRNA | BC146949.1 | - | 3, 4 | 583 | 9056877 | |
BC146966.1 | - | 3, 4 | 583 | 9056894 |
Gene Symbol: SPDYE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306141.1 | NP_001293070.1 | 4, 5 | 1097 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 4, 5 | 1097 |
Gene Symbol: SPDYE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003666.2 | 2, 3 | 388 | |||
GenBank mRNA | BC100974.2 | - | 2, 3 | 388 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 4, 5 | 986 | |||
GenBank mRNA | BC045636.1 | - | 4, 5 | 989 | ||
BC068610.1 | - | 4, 5 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
