siRNA Details
Gene Symbol: KRTAP19-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181614.1 | NP_853645.1 | 1 | 60 | ||
GenBank mRNA | AB096949.1 | - | 1 | 84 | ||
BC103836.2 | - | 1 | 60 | 40002029 | ||
BC103837.2 | - | 1 | 60 | 40002030 | ||
BC103838.1 | - | 1 | 60 | 40002031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
