siRNA Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170714.1 | NP_001164185.1 | 3 | 854 | ||
NM_001170715.1 | NP_001164186.1 | 2 | 559 | |||
NM_001170716.1 | NP_001164187.1 | 2 | 516 | |||
NM_001170717.1 | NP_001164188.1 | 2 | 578 | |||
NM_001170718.1 | NP_001164189.1 | 2 | 603 | |||
NM_001170719.1 | NP_001164190.1 | 2 | 693 | |||
NM_014567.3 | NP_055382.2 | 2 | 578 | |||
XM_005256258.4 | XP_005256315.1 | 2 | 778 | |||
XM_005256259.4 | XP_005256316.1 | 2 | 3690 | |||
XM_005256260.4 | XP_005256317.1 | 2 | 699 | |||
XM_011523475.2 | XP_011521777.1 | 2 | 727 | |||
XM_017023897.1 | XP_016879386.1 | 2 | 1285 | |||
XM_017023898.1 | XP_016879387.1 | 2 | 1285 | |||
XM_017023899.1 | XP_016879388.1 | 2 | 1285 | |||
GenBank mRNA | AB040024.1 | - | 2 | 573 | ||
AF218451.1 | - | 2 | 556 | |||
AJ242987.2 | - | 2 | 553 | |||
AK027608.1 | - | 2 | 603 | |||
AK124526.1 | - | 2 | 1998 | |||
AK124815.1 | - | 2 | 562 | |||
AK293808.1 | - | 3 | 854 | |||
AK294513.1 | - | 2 | 609 | |||
AK295809.1 | - | 2 | 488 | |||
AK314722.1 | - | 2 | 578 | |||
AY545071.1 | - | 1 | 432 | |||
BC062556.1 | - | 1 | 433 | 6427808 | ||
DC317947.1 | - | 2 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
