siRNA Details
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 8, 9 | 1274 | |||
NR_104095.1 | 7, 8 | 954 | ||||
NR_104096.1 | 7, 8 | 936 | ||||
NR_104097.1 | 7, 8 | 1047 | ||||
NR_136268.1 | 8, 9 | 1274 | ||||
GenBank mRNA | BC033195.1 | - | 3, 4 | 309 | ||
BC171827.1 | - | 7, 8 | 954 | |||
BC171832.1 | - | 7, 8 | 936 | |||
BC171834.1 | - | 7, 8 | 1208 | |||
BC171838.1 | - | 7, 8 | 1047 | |||
BC171839.1 | - | 6, 7 | 1149 | |||
DQ224421.1 | - | 8, 9 | 1175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
