siRNA Details
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 7 | 1470 | |||
GenBank mRNA | AK074778.1 | - | 7 | 1103 | ||
BC054509.1 | - | 5 | 2793 | |||
BC071951.1 | - | 7 | 1427 | |||
BC096759.1 | - | 7 | 1427 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 8 | 1235 | |||
NR_033936.3 | 7 | 1478 | ||||
NR_103496.2 | 7 | 1096 | ||||
GenBank mRNA | AF316855.1 | - | 1 | 955 | ||
AK026448.1 | - | 6 | 2094 | |||
AK309420.1 | - | 7 | 1003 | |||
AK311124.1 | - | 7 | 1275 | |||
BC007811.2 | - | 8 | 1159 | |||
BC065260.1 | - | 8 | 1152 | |||
BC072390.1 | - | 7 | 1431 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 6 | 1291 | |||
NR_102748.1 | 6 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
