siRNA Details
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 8 | 1341 | |||
NR_033936.3 | 7 | 1584 | ||||
NR_103496.2 | 7 | 1202 | ||||
GenBank mRNA | AF316855.1 | - | 1 | 1061 | ||
AK026448.1 | - | 6 | 2200 | |||
AK309420.1 | - | 7 | 1109 | |||
AK311124.1 | - | 7 | 1381 | |||
BC007811.2 | - | 8 | 1265 | |||
BC065260.1 | - | 8 | 1258 | |||
BC072390.1 | - | 7 | 1537 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 7 | 1576 | |||
GenBank mRNA | AK074778.1 | - | 7 | 1209 | ||
BC054509.1 | - | 5 | 2899 | |||
BC071951.1 | - | 7 | 1533 | |||
BC096759.1 | - | 7 | 1533 |
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049748.1 | 1 | 62 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 6 | 1397 | |||
NR_102748.1 | 6 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
