siRNA Details
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 6 | 2638 | |||
NR_102748.1 | 6 | 2343 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 8 | 2579 | |||
NR_033936.3 | 7 | 2822 | ||||
NR_103496.2 | 7 | 2440 | ||||
GenBank mRNA | AF316855.1 | - | 3 | 2238 | ||
AK026448.1 | - | 7 | 2494 | |||
BC007811.2 | - | 9 | 2505 | |||
BC065260.1 | - | 8 | 2496 | |||
BC072390.1 | - | 7 | 2779 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 7 | 2840 | |||
GenBank mRNA | AK074778.1 | - | 7 | 2468 | ||
BC054509.1 | - | 5 | 4156 | |||
BC071951.1 | - | 7 | 2792 | |||
BC096759.1 | - | 7 | 2792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
