siRNA Details
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101155.1 | - | 3 | 343 | 40015679 |
Gene Symbol: TAS2R19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF116728.1 | - | 1 | 1272 |
Gene Symbol: TAS2R30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097643.1 | NP_001091112.1 | 1 | 343 |
Gene Symbol: TAS2R31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176885.2 | NP_795366.2 | 1 | 415 | ||
GenBank mRNA | BC117421.1 | - | 1 | 416 | 40125972 |
Gene Symbol: TAS2R45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176886.2 | NP_795367.2 | 3 | 343 | ||
GenBank mRNA | BC101154.1 | - | 3 | 343 | 40015678 | |
BC101156.1 | - | 3 | 343 | 40015680 |
Gene Symbol: TAS2R46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176887.2 | NP_795368.2 | 1 | 343 | ||
GenBank mRNA | BC115401.2 | - | 1 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
