siRNA Details
Gene Symbol: OR4F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004195.2 | NP_001004195.2 | 1 | 44 | ||
GenBank mRNA | AY972819.1 | - | 1 | 110 | ||
BC136907.1 | - | 1 | 80 | 9020898 | ||
BC136908.1 | - | 1 | 80 | 9020899 |
Gene Symbol: OR4F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005484.1 | NP_001005484.1 | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
