siRNA Details
Gene Symbol: ZNF767P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027788.1 | 2 | 326 | |||
NR_027789.1 | 2 | 326 | ||||
GenBank mRNA | AK022762.1 | - | 2 | 263 | ||
AK295771.1 | - | 2 | 164 | |||
BC047675.1 | - | 2 | 288 | 5727587 | ||
DA249066.1 | - | 2 | 218 | |||
DA418417.1 | - | 2 | 237 | |||
DC295958.1 | - | 2 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
