siRNA Details
Gene Symbol: LOC554249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126049.1 | 2 | 414 | |||
GenBank mRNA | BC084570.1 | - | 2 | 328 | 6378300 |
Gene Symbol: GLIDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015363.2 | 2 | 492 | |||
NR_126044.1 | 2 | 474 | ||||
NR_126045.1 | 2 | 474 | ||||
NR_126046.1 | 1 | 589 | ||||
GenBank mRNA | AK092305.1 | - | 2 | 391 | ||
AK095690.1 | - | 2 | 375 | |||
BC019880.1 | - | 1 | 100 | |||
BC027338.1 | - | 2 | 378 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126048.1 | 2 | 418 | |||
GenBank mRNA | AK292642.1 | - | 2 | 345 |
Gene Symbol: LOC103908605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA585556.1 | - | 2 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
