siRNA Details
Gene Symbol: LOC103908605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126050.1 | 1 | 260 | |||
GenBank mRNA | DA585556.1 | - | 1 | 260 | ||
DR003081.1 | - | 207 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292642.1 | - | 1 | 239 |
Gene Symbol: LOC103908604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA708908.1 | - | 1 | 66 | ||
DA742734.1 | - | 1 | 287 |
Gene Symbol: LOC554249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC084570.1 | - | 1 | 222 | 6378300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
