siRNA Details
Gene Symbol: NUTM2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017561.1 | NP_060031.1 | 5 | 1280 | ||
GenBank mRNA | AL133071.1 | - | 4 | 833 | ||
BC130390.1 | - | 5 | 1280 | 40146421 | ||
BC143798.1 | - | 5 | 1280 | 9052312 |
Gene Symbol: NUTM2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045477.2 | NP_001038942.1 | 5 | 1235 | ||
NM_001170741.1 | NP_001164212.1 | 5 | 1235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
