siRNA Details
Gene Symbol: LCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178356.3 | NP_848133.1 | 1 | 107 | ||
GenBank mRNA | BC113446.1 | - | 1 | 107 | 8322498 | |
BC113448.1 | - | 1 | 107 | 8322500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
