siRNA Details
Gene Symbol: LCE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178428.3 | NP_848515.1 | 2 | 127 | ||
GenBank mRNA | BC119707.2 | - | 2 | 127 | 40117159 | |
BC127647.1 | - | 2 | 127 | |||
BG217525.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
