siRNA Details
Gene Symbol: LCE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178428.3 | NP_848515.1 | 2 | 71 | ||
GenBank mRNA | BC119707.2 | - | 2 | 71 | 40117159 | |
BC127647.1 | - | 2 | 71 | |||
BG217525.1 | - | 1 | 45 |
Gene Symbol: LCE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014357.4 | NP_055172.1 | 2 | 74 | ||
GenBank mRNA | AF005080.1 | - | 2 | 71 | ||
AF086289.1 | - | 2 | 74 | |||
BC117235.1 | - | 2 | 65 | |||
BC117237.1 | - | 2 | 65 |
Gene Symbol: LCE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178429.3 | NP_848516.1 | 1 | 55 | ||
GenBank mRNA | BC125197.1 | - | 1 | 55 |
Gene Symbol: LCE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178430.3 | NP_848517.1 | 2 | 60 | ||
GenBank mRNA | BC130603.1 | - | 2 | 60 | 40146634 | |
BC130605.1 | - | 2 | 60 | 40146636 |
Gene Symbol: LCE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178434.2 | NP_848521.1 | 1 | 90 | ||
GenBank mRNA | BC130365.1 | - | 1 | 90 | 40146396 | |
BC130367.1 | - | 1 | 90 | 40146398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
