siRNA Details
Gene Symbol: LCE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178430.3 | NP_848517.1 | 2 | 92 | ||
GenBank mRNA | BC130603.1 | - | 2 | 92 | 40146634 | |
BC130605.1 | - | 2 | 92 | 40146636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
