siRNA Details
Gene Symbol: LCE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178434.2 | NP_848521.1 | 1 | 122 | ||
GenBank mRNA | BC130365.1 | - | 1 | 122 | 40146396 | |
BC130367.1 | - | 1 | 122 | 40146398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
