siRNA Details
Gene Symbol: LCE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032563.1 | NP_115952.1 | 2 | 77 | ||
GenBank mRNA | AB048288.1 | - | 2 | 77 | ||
BC069291.1 | - | 2 | 70 | 7262717 | ||
BC101784.1 | - | 2 | 60 | 8069290 | ||
BC120935.1 | - | 1 | 20 | 40115032 | ||
BC120936.1 | - | 1 | 18 |
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 496 | ||
GenBank mRNA | BC031811.1 | - | 1 | 496 | ||
BC104232.1 | - | 1 | 104 | 40032247 | ||
BC104233.1 | - | 1 | 106 | 40032251 | ||
BC133657.1 | - | 1 | 106 |
Gene Symbol: LCE3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178431.1 | NP_848518.1 | 1 | 20 | ||
GenBank mRNA | BC120933.1 | - | 1 | 20 | ||
BC120934.1 | - | 1 | 20 |
Gene Symbol: LCE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178433.1 | NP_848520.1 | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
