siRNA Details
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 2 | 236 | |||
NR_029426.1 | 5 | 528 | ||||
GenBank mRNA | AK097091.1 | - | 6 | 908 | ||
AK289832.1 | - | 5 | 610 | |||
AK315849.1 | - | 5 | 601 | |||
BC131740.1 | - | 3 | 342 | |||
X75940.1 | - | 2 | 237 | |||
X83300.1 | - | 3 | 351 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 5 | 491 | |||
GenBank mRNA | AK290693.1 | - | 5 | 458 | ||
BC147000.1 | - | 5 | 445 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 5 | 632 | |||
GenBank mRNA | BC034017.1 | - | 5 | 633 | ||
BC067351.1 | - | 3 | 330 | |||
BC140728.1 | - | 4 | 455 | |||
BC171739.1 | - | 4 | 455 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027386.2 | 5 | 610 | |||
GenBank mRNA | AK289965.1 | - | 4 | 582 | ||
BT006760.1 | - | 3 | 285 | |||
DT218991.1 | - | 5 | 530 | |||
U21555.1 | - | LIT2100 5G4 pseudogene |
2 | 73 | ||
X83299.1 | - | 5 | 561 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 3 | 279 | |||
NR_133000.1 | 3 | 279 | ||||
GenBank mRNA | AW955360.1 | - | 2 | 107 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 5 | 519 | |||
NR_132997.1 | 7 | 778 | ||||
NR_132998.1 | 4 | 446 | ||||
GenBank mRNA | BC144584.1 | - | 5 | 455 | ||
BC144585.1 | - | 4 | 382 | |||
DA269273.1 | - | 4 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
