siRNA Details
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184825.1 | NP_001171754.1 | 2 | 256 | ||
NM_001184826.1 | NP_001171755.1 | 2 | 256 | |||
NM_001297773.1 | NP_001284702.1 | 2 | 204 | |||
NM_006905.2 | NP_008836.2 | 2 | 256 | |||
XM_005259065.3 | XP_005259122.1 | 2 | 255 | |||
XM_011527125.1 | XP_011525427.1 | 2 | 255 | |||
XM_017026994.1 | XP_016882483.1 | 2 | 255 | |||
GenBank mRNA | BC022338.1 | - | 2 | 256 | 4732357 | |
BC058285.1 | - | 2 | 180 | 4770394 | ||
CR450329.1 | - | 2 | 118 | |||
J04539.1 | - | 2 | 178 | |||
M20879.1 | - | 2 | 204 | |||
M20881.1 | - | 2 | 235 | |||
M21822.1 | - | 2 | 191 | |||
M25385.1 | - | 2 | 154 | |||
M37397.1 | - | 2 | 208 |
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 2 | 250 | ||
XM_011527126.1 | XP_011525428.1 | 2 | 250 | |||
XM_011527127.1 | XP_011525429.1 | 2 | 250 | |||
GenBank mRNA | BC005924.1 | - | 2 | 215 | 4043628 | |
DA833589.1 | - | 2 | 250 | |||
M23575.1 | - | 2 | 349 | |||
M30628.1 | - | 3 | 99 | |||
M34420.1 | - | 2 | 199 | |||
M37399.1 | - | 2 | 204 | |||
M93061.1 | - | 2 | 176 | |||
S59493.1 | - | 2 | 176 | |||
S59494.1 | - | 3 | 105 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008405.1 | - | 2 | 215 | 4043484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
