siRNA Details
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184825.1 | NP_001171754.1 | 1, 2 | 208 | ||
NM_001184826.1 | NP_001171755.1 | 1, 2 | 208 | |||
NM_001297773.1 | NP_001284702.1 | 1, 2 | 156 | |||
NM_006905.2 | NP_008836.2 | 1, 2 | 208 | |||
XM_005259065.3 | XP_005259122.1 | 1, 2 | 207 | |||
XM_011527125.1 | XP_011525427.1 | 1, 2 | 207 | |||
XM_017026994.1 | XP_016882483.1 | 1, 2 | 207 | |||
GenBank mRNA | AK300152.1 | - | 1, 2 | 163 | ||
AK300206.1 | - | 1, 2 | 201 | |||
AK311491.1 | - | 1, 2 | 169 | |||
BC022338.1 | - | 1, 2 | 208 | 4732357 | ||
BC058285.1 | - | 1, 2 | 132 | 4770394 | ||
CR450329.1 | - | 1, 2 | 70 | |||
J04539.1 | - | 1, 2 | 130 | |||
M17908.1 | - | 1, 2 | 136 | |||
M20879.1 | - | 1, 2 | 156 | |||
M20881.1 | - | 1, 2 | 187 | |||
M21822.1 | - | 1, 2 | 143 | |||
M25385.1 | - | 1, 2 | 106 | |||
M33663.1 | - | 1, 2 | 133 | |||
M33664.1 | - | 1, 2 | 70 | |||
M34715.1 | - | 1, 2 | 231 | |||
M37397.1 | - | 1, 2 | 160 |
Gene Symbol: PSG10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026824.1 | 1, 2 | 169 | |||
GenBank mRNA | AK309876.1 | - | 1, 2 | 169 |
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 1, 2 | 242 | ||
GenBank mRNA | BC022316.1 | - | 1, 2 | 169 | 4731250 | |
M20882.1 | - | 1, 2 | 153 |
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M23575.1 | - | 1, 2 | 301 | ||
M34420.1 | - | 1, 2 | 151 | |||
M37399.1 | - | 1, 2 | 156 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 1, 2 | 308 | ||
NM_002780.4 | NP_002771.2 | 1, 2 | 308 | |||
NM_213633.2 | NP_998798.1 | 1, 2 | 308 | |||
XM_017026997.1 | XP_016882486.1 | 1, 2 | 202 | |||
XM_017027001.1 | XP_016882490.1 | 1, 2 | 202 | |||
XM_017027002.1 | XP_016882491.1 | 1, 2 | 202 | |||
GenBank mRNA | AK075149.1 | - | 2, 3 | 202 | ||
BC008405.1 | - | 1, 2 | 167 | 4043484 | ||
BC063127.1 | - | 1, 2 | 176 | 4768860 | ||
BX403624.2 | - | 1, 2 | 160 | |||
DA837940.1 | - | 1, 2 | 308 | |||
M94891.1 | - | 1, 2 | 156 | |||
U18468.1 | - | 1, 2 | 75 | |||
U18469.1 | - | 1, 2 | 97 | |||
X17097.1 | - | 1, 2 | 162 |
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130014.1 | NP_001123486.1 | 1, 2 | 201 | ||
NM_002781.3 | NP_002772.3 | 1, 2 | 201 | |||
XM_011527132.2 | XP_011525434.1 | 1, 2 | 199 | |||
XM_017027003.1 | XP_016882492.1 | 1, 2 | 199 | |||
GenBank mRNA | BC012607.1 | - | 1, 2 | 168 | 4043912 | |
BG622527.1 | - | 1, 2 | 169 | |||
DA835167.1 | - | 1, 2 | 201 | |||
M25384.1 | - | 1, 2 | 169 | |||
M73713.1 | - | 1, 2 | 155 |
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031850.3 | NP_001027020.1 | 1, 2 | 202 | ||
NM_002782.4 | NP_002773.1 | 1, 2 | 202 | |||
GenBank mRNA | BC020652.1 | - | 1, 2 | 169 | 4770562 | |
DA833634.1 | - | 1, 2 | 202 | |||
M31125.1 | - | 1, 2 | 115 | |||
M33666.1 | - | 1, 2 | 122 | |||
X17098.1 | - | 1, 2 | 123 |
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290042.1 | NP_001276971.1 | 1, 2 | 172 | ||
NM_002783.2 | NP_002774.2 | 1, 2 | 172 | |||
GenBank mRNA | BC030979.1 | - | 1, 2 | 167 | ||
BG434892.1 | - | 1, 2 | 168 | |||
BP321474.1 | - | 1, 2 | 171 | |||
BX952498.1 | - | 1, 2 | 170 | |||
U18467.1 | - | 1, 2 | 122 |
Gene Symbol: PSG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130167.1 | NP_001123639.1 | 1, 2 | 168 | ||
NM_182707.2 | NP_874366.1 | 1, 2 | 168 | |||
XM_011526986.1 | XP_011525288.1 | 1, 2 | 181 | |||
GenBank mRNA | BC137500.1 | - | 1, 2 | 168 | 9021500 | |
BC142628.1 | - | 1, 2 | 156 | 40147510 | ||
CR749812.1 | - | 1, 2 | 167 |
Gene Symbol: PSG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301707.1 | NP_001288636.1 | 1, 2 | 202 | ||
NM_001301708.1 | NP_001288637.1 | 1, 2 | 202 | |||
NM_001301709.1 | NP_001288638.1 | 1, 2 | 202 | |||
NM_002784.4 | NP_002775.3 | 1, 2 | 202 | |||
XM_005259075.3 | XP_005259132.1 | 1, 2 | 164 | |||
XM_017027004.1 | XP_016882493.1 | 1, 2 | 164 | |||
XM_017027005.1 | XP_016882494.1 | 1, 2 | 197 | |||
XM_017027006.1 | XP_016882495.1 | 1, 2 | 197 | |||
XM_017027007.1 | XP_016882496.1 | 1, 2 | 164 | |||
GenBank mRNA | AK300187.1 | - | 1, 3 | 165 | ||
AK300229.1 | - | 1, 2 | 167 | |||
AK313256.1 | - | 1, 2 | 169 | |||
BC005925.1 | - | 1, 2 | 167 | 4044135 | ||
BC020759.1 | - | 1, 2 | 167 | 4731739 | ||
BX952516.1 | - | 1, 2 | 170 | |||
CR407605.1 | - | 1, 2 | 70 | |||
DC380417.1 | - | 1, 2 | 202 | |||
M34421.1 | - | 1, 2 | 148 | |||
M34481.1 | - | 1, 2 | 139 | |||
M58591.1 | - | 1, 2 | 141 | |||
M94890.1 | - | 1, 2 | 137 | |||
U25987.1 | - | 1, 2 | 80 | |||
X17610.1 | - | 1, 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
