siRNA Details
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123363.3 | NP_001116835.1 | 21 | 4012 | ||
XM_011511766.2 | XP_011510068.1 | 20 | 3596 | |||
XM_011511767.2 | XP_011510069.1 | 20 | 3596 | |||
XM_011511769.2 | XP_011510071.1 | 20 | 3596 | |||
XM_011511770.2 | XP_011510072.1 | 20 | 3596 | |||
XM_017004839.1 | XP_016860328.1 | 20 | 3596 | |||
XM_017004840.1 | XP_016860329.1 | 20 | 3669 | |||
XM_017004841.1 | XP_016860330.1 | 20 | 3799 | |||
XM_017004842.1 | XP_016860331.1 | 20 | 3576 | |||
XM_017004843.1 | XP_016860332.1 | 20 | 3596 | |||
XM_017004844.1 | XP_016860333.1 | 18 | 3258 |
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 20 | 6533 | ||
XM_005264002.2 | XP_005264059.1 | 20 | 6568 | |||
XM_005264003.2 | XP_005264060.1 | 20 | 6496 | |||
XM_005264004.2 | XP_005264061.1 | 20 | 6496 | |||
XM_005264005.4 | XP_005264062.1 | 20 | 6556 | |||
XM_005264007.2 | XP_005264064.1 | 20 | 3568 | |||
XM_011511575.2 | XP_011509877.1 | 20 | 6471 | |||
XM_011511576.2 | XP_011509878.1 | 21 | 6319 | |||
XM_011511578.2 | XP_011509880.1 | 20 | 3610 | |||
XM_017004623.1 | XP_016860112.1 | 20 | 6496 | |||
GenBank mRNA | AB209483.1 | - | 17 | 6149 | ||
AK025711.1 | - | 1 | 709 | |||
D42063.1 | - | 20 | 6534 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 20 | 3450 | ||
XM_011532845.2 | XP_011531147.1 | 21 | 3649 | |||
XM_011532849.2 | XP_011531151.1 | 21 | 3649 | |||
XM_011532850.1 | XP_011531152.1 | 21 | 3649 | |||
XM_011532851.2 | XP_011531153.1 | 21 | 3649 | |||
XM_011532852.2 | XP_011531154.1 | 21 | 3649 | |||
XM_017004103.1 | XP_016859592.1 | 21 | 3649 | |||
XM_017004104.1 | XP_016859593.1 | 20 | 3766 | |||
XM_017004105.1 | XP_016859594.1 | 20 | 3614 | |||
XM_017004106.1 | XP_016859595.1 | 20 | 3733 | |||
XM_017004107.1 | XP_016859596.1 | 21 | 3649 | |||
GenBank mRNA | AK303443.1 | - | 6 | 1366 | ||
CR749330.1 | - | 20 | 3450 | |||
DC353941.1 | - | 1 | 31 |
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 20 | 3496 | ||
XM_017004845.1 | XP_016860334.1 | 24 | 3947 | |||
XM_017004846.1 | XP_016860335.1 | 20 | 3618 | |||
XM_017004847.1 | XP_016860336.1 | 20 | 3615 | |||
XM_017004848.1 | XP_016860337.1 | 24 | 3947 | |||
XM_017004849.1 | XP_016860338.1 | 24 | 3947 | |||
XM_017004850.1 | XP_016860339.1 | 24 | 3947 | |||
GenBank mRNA | AK300276.1 | - | 1 | 283 | ||
AK300352.1 | - | 9 | 1836 | |||
AK302429.1 | - | 20 | 3496 | |||
AY354203.1 | - | 7 | 1472 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 20 | 3539 | ||
XM_017004738.1 | XP_016860227.1 | 21 | 3588 | |||
XM_017004739.1 | XP_016860228.1 | 21 | 3588 | |||
GenBank mRNA | AK310529.1 | - | 6 | 1318 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 20 | 3564 | ||
XM_011511017.2 | XP_011509319.1 | 20 | 3511 | |||
XM_011511018.2 | XP_011509320.1 | 20 | 3514 | |||
XM_011511019.2 | XP_011509321.1 | 20 | 3510 | |||
XM_011511020.2 | XP_011509322.1 | 20 | 3510 | |||
GenBank mRNA | AK302921.1 | - | 16 | 2872 | ||
BX537861.1 | - | 15 | 4463 |
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 20 | 3556 | ||
XM_006712794.3 | XP_006712857.2 | 20 | 3587 | |||
XM_017005080.1 | XP_016860569.1 | 20 | 3576 | |||
XM_017005081.1 | XP_016860570.1 | 20 | 3587 | |||
XM_017005082.1 | XP_016860571.1 | 18 | 3258 | |||
GenBank mRNA | AB209082.1 | - | 1 | 455 | ||
EU794682.1 | - | 20 | 3556 | |||
U64675.2 | - | 20 | 3556 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 20 | 3671 | ||
XM_005263747.4 | XP_005263804.1 | 20 | 3669 | |||
XM_011511733.2 | XP_011510035.1 | 20 | 3669 | |||
XM_017004825.1 | XP_016860314.1 | 20 | 3669 | |||
GenBank mRNA | AF012086.1 | - | 1 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
