siRNA Details
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 20 | 6676 | ||
XM_005264002.2 | XP_005264059.1 | 20 | 6711 | |||
XM_005264003.2 | XP_005264060.1 | 20 | 6639 | |||
XM_005264004.2 | XP_005264061.1 | 20 | 6639 | |||
XM_005264005.4 | XP_005264062.1 | 20 | 6699 | |||
XM_005264007.2 | XP_005264064.1 | 20 | 3711 | |||
XM_011511575.2 | XP_011509877.1 | 20 | 6614 | |||
XM_011511576.2 | XP_011509878.1 | 21 | 6462 | |||
XM_011511578.2 | XP_011509880.1 | 20 | 3753 | |||
XM_017004623.1 | XP_016860112.1 | 20 | 6639 | |||
GenBank mRNA | AB209483.1 | - | 17 | 6292 | ||
AK025711.1 | - | 1 | 852 | |||
D42063.1 | - | 20 | 6677 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 20 | 3682 | ||
XM_017004738.1 | XP_016860227.1 | 21 | 3731 | |||
XM_017004739.1 | XP_016860228.1 | 21 | 3731 | |||
GenBank mRNA | AK310529.1 | - | 6 | 1461 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 20 | 3707 | ||
XM_011511017.2 | XP_011509319.1 | 20 | 3654 | |||
XM_011511018.2 | XP_011509320.1 | 20 | 3657 | |||
XM_011511019.2 | XP_011509321.1 | 20 | 3653 | |||
XM_011511020.2 | XP_011509322.1 | 20 | 3653 | |||
GenBank mRNA | BX537861.1 | - | 15 | 4606 |
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 20 | 3699 | ||
XM_006712794.3 | XP_006712857.2 | 20 | 3730 | |||
XM_017005080.1 | XP_016860569.1 | 20 | 3719 | |||
XM_017005081.1 | XP_016860570.1 | 20 | 3730 | |||
XM_017005082.1 | XP_016860571.1 | 18 | 3401 | |||
GenBank mRNA | AB209082.1 | - | 1 | 598 | ||
EU794682.1 | - | 20 | 3699 | |||
U64675.2 | - | 20 | 3699 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123363.3 | NP_001116835.1 | 21 | 4155 | ||
XM_011511766.2 | XP_011510068.1 | 20 | 3739 | |||
XM_011511767.2 | XP_011510069.1 | 20 | 3739 | |||
XM_011511769.2 | XP_011510071.1 | 20 | 3739 | |||
XM_011511770.2 | XP_011510072.1 | 20 | 3739 | |||
XM_017004839.1 | XP_016860328.1 | 20 | 3739 | |||
XM_017004840.1 | XP_016860329.1 | 20 | 3812 | |||
XM_017004841.1 | XP_016860330.1 | 20 | 3942 | |||
XM_017004842.1 | XP_016860331.1 | 20 | 3719 | |||
XM_017004843.1 | XP_016860332.1 | 20 | 3739 | |||
XM_017004844.1 | XP_016860333.1 | 18 | 3401 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 20 | 3814 | ||
XM_005263747.4 | XP_005263804.1 | 20 | 3812 | |||
XM_011511733.2 | XP_011510035.1 | 20 | 3812 | |||
XM_017004825.1 | XP_016860314.1 | 20 | 3812 | |||
GenBank mRNA | AF012086.1 | - | 1 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
