siRNA Details
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 4 | 479 | ||
XM_005273335.2 | XP_005273392.1 | 2 | 320 | |||
XM_006711592.2 | XP_006711655.1 | 3 | 1267 | |||
XM_011510072.2 | XP_011508374.1 | 3 | 319 | |||
XM_011510073.2 | XP_011508375.1 | 3 | 797 | |||
XM_011510074.2 | XP_011508376.1 | 3 | 338 | |||
XM_011510075.2 | XP_011508377.1 | 3 | 825 | |||
XM_011510077.2 | XP_011508379.1 | 2 | 261 | |||
XM_017002611.1 | XP_016858100.1 | 2 | 735 | |||
GenBank mRNA | AK098669.1 | - | 2 | 213 | ||
AK125728.1 | - | 2 | 317 | |||
BC007734.2 | - | 2 | 300 | |||
BC107762.1 | - | 2 | 304 | |||
DA756409.1 | - | 4 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
