siRNA Details
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 4 | 593 | ||
XM_005273335.2 | XP_005273392.1 | 2 | 434 | |||
XM_006711592.2 | XP_006711655.1 | 3 | 1381 | |||
XM_011510072.2 | XP_011508374.1 | 3 | 433 | |||
XM_011510073.2 | XP_011508375.1 | 3 | 911 | |||
XM_011510074.2 | XP_011508376.1 | 3 | 452 | |||
XM_011510075.2 | XP_011508377.1 | 3 | 939 | |||
XM_011510077.2 | XP_011508379.1 | 2 | 375 | |||
XM_017002611.1 | XP_016858100.1 | 2 | 849 | |||
XM_017002613.1 | XP_016858102.1 | 1 | 60 | |||
GenBank mRNA | AK098669.1 | - | 2 | 327 | ||
AK125728.1 | - | 2 | 431 | |||
BC007734.2 | - | 2 | 414 | |||
BC107762.1 | - | 2 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
