siRNA Details
Gene Symbol: ANAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511634.2 | XP_011509936.1 | 2 | 492 | ||
XM_017004712.1 | XP_016860201.1 | 2 | 492 | |||
XM_017004713.1 | XP_016860202.1 | 2 | 492 | |||
XM_017004714.1 | XP_016860203.1 | 2 | 440 | |||
GenBank mRNA | BX282336.1 | - | 2 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
