siRNA Details
Gene Symbol: ANAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022662.3 | NP_073153.1 | 2 | 263 | ||
XM_011511633.2 | XP_011509935.1 | 2 | 295 | |||
XM_011511634.2 | XP_011509936.1 | 2 | 290 | |||
XM_011511636.2 | XP_011509938.1 | 2 | 295 | |||
XM_011511637.2 | XP_011509939.1 | 2 | 295 | |||
XM_017004710.1 | XP_016860199.1 | 2 | 295 | |||
XM_017004711.1 | XP_016860200.1 | 2 | 295 | |||
XM_017004712.1 | XP_016860201.1 | 2 | 290 | |||
XM_017004713.1 | XP_016860202.1 | 2 | 290 | |||
XM_017004714.1 | XP_016860203.1 | 2 | 238 | |||
XM_017004715.1 | XP_016860204.1 | 2 | 295 | |||
GenBank mRNA | AJ278357.1 | - | 2 | 278 | ||
BC104902.1 | - | 2 | 65 | 8143905 | ||
BC104904.1 | - | 2 | 65 | 8143907 | ||
BX282336.1 | - | 2 | 263 |
Gene Symbol: ANAPC1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037931.1 | 2 | 138 | |||
GenBank mRNA | AK310642.1 | - | 2 | 138 |
Gene Symbol: LOC107985931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005472.1 | XP_016860961.1 | 3 | 231 | ||
XM_017005473.1 | XP_016860962.1 | 3 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
