siRNA Details
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000972.2 | NP_000963.1 | 3, 4 | 302 | ||
GenBank mRNA | AK222534.1 | - | 3, 4 | 299 | ||
AK291123.1 | - | 3, 4 | 294 | |||
AK311743.1 | - | 3, 4 | 298 | |||
BC005128.1 | - | 1 | 285 | 3938260 | ||
BC021052.1 | - | 1 | 276 | |||
BC021979.2 | - | 1 | 274 | 4053567 | ||
BC023594.2 | - | 3, 4 | 333 | 4841982 | ||
BC023624.2 | - | 3, 4 | 338 | 4810336 | ||
BC032533.1 | - | 1 | 321 | |||
BC071900.1 | - | 1 | 274 | 6090366 | ||
BC071901.1 | - | 2, 3 | 282 | 6262332 | ||
BC073802.1 | - | 1 | 273 | 4336858 | ||
BC105290.1 | - | 1 | 299 | 6495031 | ||
M36072.1 | - | 3, 4 | 303 | |||
X06705.1 | - | 3, 4 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
