siRNA Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006.4 | NP_000997.1 | 6 | 859 | ||
GenBank mRNA | AK129825.1 | - | 6 | 802 | ||
AK311788.1 | - | 6 | 802 | |||
BC000204.1 | - | 1 | 791 | 3350750 | ||
BC001708.1 | - | 6 | 821 | 3544072 | ||
BC004981.1 | - | 6 | 791 | 2906117 | ||
BC006298.2 | - | 1 | 784 | 4079651 | ||
BC009219.2 | - | 1 | 791 | 3927639 | ||
BC009404.2 | - | 1 | 793 | 4308955 | ||
BC017123.2 | - | 1 | 791 | 3922810 | ||
BC018954.2 | - | 1 | 793 | |||
BC019072.2 | - | 1 | 842 | 5020581 | ||
BC030161.2 | - | 1 | 842 | 5020904 | ||
BC066926.1 | - | 1 | 803 | 4826149 | ||
BC070211.1 | - | 1 | 804 | 5809691 | ||
BC071916.1 | - | 1 | 791 | 6285242 | ||
L13802.1 | - | 6 | 785 | |||
M77234.1 | - | 6 | 796 | |||
M84711.1 | - | 6 | 815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
