siRNA Details
Gene Symbol: SRSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000100.1 | XP_016855589.1 | 3 | 4108 | ||
XM_017000101.1 | XP_016855590.1 | 4 | 3651 | |||
XM_017000102.1 | XP_016855591.1 | 4 | 1278 | |||
GenBank mRNA | AK055311.1 | - | 1 | 1190 | ||
AK125834.1 | - | 1 | 2585 | |||
BC071575.1 | - | 3 | 3509 | |||
BX648939.1 | - | 1 | 2441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
