siRNA Details
Gene Symbol: GAL3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022134.2 | NP_071417.2 | 4 | 577 | ||
GenBank mRNA | AB040610.1 | - | 4 | 578 | ||
BC117293.1 | - | 4 | 495 | 40125844 | ||
BC117295.1 | - | 4 | 495 | 40125846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
