siRNA Details
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 2 | 718 | ||
XM_017012888.1 | XP_016868377.1 | 2 | 718 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 2 | 714 | ||
XM_017012933.1 | XP_016868422.1 | 2 | 718 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 3 | 755 | |||
GenBank mRNA | BC045636.1 | - | 3 | 758 | ||
BC068610.1 | - | 3 | 758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
