siRNA Details
Gene Symbol: KRTAP10-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198694.3 | NP_941967.3 | 1 | 231 | ||
GenBank mRNA | AB076352.1 | - | 1 | 231 | ||
BC120949.1 | - | 1 | 230 | 40115143 | ||
BC120950.1 | - | 1 | 230 | 40115146 |
Gene Symbol: KRTAP10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198691.2 | NP_941964.2 | 1 | 268 | ||
GenBank mRNA | AB076347.1 | - | 1 | 268 | ||
BC120959.2 | - | 1 | 262 | 40115271 | ||
BC120960.1 | - | 1 | 262 | 40115274 |
Gene Symbol: KRTAP10-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198699.1 | NP_941972.1 | 1 | 268 | ||
GenBank mRNA | AB076364.1 | - | 1 | 268 | ||
BC137339.1 | - | 1 | 268 | 9021337 | ||
BC137341.1 | - | 1 | 268 | 9021339 |
Gene Symbol: KRTAP10-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198693.3 | NP_941966.1 | 1 | 270 | ||
GenBank mRNA | AB076348.1 | - | 1 | 270 |
Gene Symbol: KRTAP10-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198696.2 | NP_941969.2 | 1 | 268 | ||
GenBank mRNA | AB076350.1 | - | 1 | 268 | ||
BC133677.1 | - | 1 | 247 | 40119288 |
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 283 | ||
GenBank mRNA | AB076351.1 | - | 1 | 283 | ||
BC125048.1 | - | 1 | 272 | 40116133 | ||
BC125049.1 | - | 1 | 272 | 40116140 |
Gene Symbol: KRTAP10-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198688.3 | NP_941961.3 | 1 | 274 | ||
GenBank mRNA | AB076353.1 | - | 1 | 274 | ||
BC140725.1 | - | 287 | 9021593 |
Gene Symbol: KRTAP10-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198689.2 | NP_941962.1 | 1 | 278 | ||
GenBank mRNA | AB076354.1 | - | 1 | 278 | ||
BC119643.2 | - | 1 | 275 | 40115724 |
Gene Symbol: KRTAP10-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198690.2 | NP_941963.2 | 1 | 272 | ||
GenBank mRNA | AB076356.1 | - | 1 | 272 | ||
BC131613.1 | - | 1 | 248 | 40115414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
