siRNA Details
Gene Symbol: KRTAP10-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198699.1 | NP_941972.1 | 1 | 311 | ||
GenBank mRNA | AB076364.1 | - | 1 | 311 | ||
BC137339.1 | - | 1 | 311 | 9021337 | ||
BC137341.1 | - | 1 | 311 | 9021339 |
Gene Symbol: KRTAP10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198691.2 | NP_941964.2 | 1 | 311 | ||
GenBank mRNA | AB076347.1 | - | 1 | 311 | ||
BC120959.2 | - | 1 | 305 | 40115271 | ||
BC120960.1 | - | 1 | 305 | 40115274 |
Gene Symbol: KRTAP10-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198692.2 | NP_941965.2 | 1 | 311 | ||
GenBank mRNA | AB076359.1 | - | 1 | 311 | ||
BC131611.1 | - | 1 | 292 | 40115290 |
Gene Symbol: KRTAP10-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198693.3 | NP_941966.1 | 1 | 313 | ||
GenBank mRNA | AB076348.1 | - | 1 | 313 |
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 683 | ||
GenBank mRNA | AB076351.1 | - | 1 | 683 | ||
BC125048.1 | - | 1 | 672 | 40116133 | ||
BC125049.1 | - | 1 | 672 | 40116140 |
Gene Symbol: KRTAP10-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198695.2 | NP_941968.2 | 1 | 345 | ||
GenBank mRNA | AB076355.1 | - | 1 | 345 | ||
BC127016.1 | - | 1 | 339 | 40119338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
