siRNA Details
Gene Symbol: TAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201536.1 | NP_001188465.1 | 4, 5 | 616 | ||
NM_005681.3 | NP_005672.1 | 4, 5 | 616 | |||
NM_139352.2 | NP_647603.1 | 3, 4 | 446 | |||
XM_005273343.3 | XP_005273400.1 | 4, 5 | 779 | |||
XM_006711613.3 | XP_006711676.1 | 4, 5 | 629 | |||
XM_017002759.1 | XP_016858248.1 | 4, 5 | 585 | |||
XM_017002760.1 | XP_016858249.1 | 3, 4 | 393 | |||
GenBank mRNA | AK001054.1 | - | 3, 4 | 298 | ||
AK290970.1 | - | 4, 5 | 561 | |||
AK299536.1 | - | 4, 5 | 616 | |||
AK315740.1 | - | 4, 5 | 600 | |||
BC013808.1 | - | 4, 5 | 596 | 3918866 | ||
L39060.1 | - | 3, 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
