siRNA Details
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 2 | 189 | ||
XM_017022481.1 | XP_016877970.1 | 2 | 189 | |||
XM_017022482.1 | XP_016877971.1 | 2 | 189 | |||
GenBank mRNA | DB040608.1 | - | 2 | 189 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 2 | 227 | ||
NM_001322400.1 | NP_001309329.1 | 2 | 227 | |||
GenBank mRNA | AK302092.1 | - | 2 | 189 | ||
CR936602.1 | - | 2 | 190 | |||
DB042163.1 | - | 2 | 187 | |||
DB044490.1 | - | 2 | 227 |
Gene Symbol: GOLGA6L17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111962.1 | 2 | 191 | |||
GenBank mRNA | AK301885.1 | - | 191 |
Gene Symbol: GOLGA6L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310153.1 | NP_001297082.1 | 2 | 189 | ||
GenBank mRNA | AK301968.1 | - | 2 | 156 |
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 2 | 189 | |||
GenBank mRNA | AK097432.1 | - | 2 | 191 | ||
AK130562.1 | - | 2 | 282 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 2 | 155 | ||
NM_198181.3 | NP_937824.3 | 2 | 227 | |||
XM_011521601.2 | XP_011519903.1 | 2 | 838 | |||
XM_017022226.1 | XP_016877715.1 | 2 | 844 | |||
GenBank mRNA | BC041854.1 | - | 2 | 155 | 5270411 | |
CR936812.1 | - | 1 | 1824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
